Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat alpha-Actinin 1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF8279
This item is not returnable.
View return policy
Description
alpha-Actinin 1 Polyclonal specifically detects alpha-Actinin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
alpha-Actinin 1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
actinin 1 smooth muscle, actinin, alpha 1, ACTN1, alphaActinin 1, alpha-Actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, BDPLT15, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1 | |
Goat | |
Affinity Purified | |
RUO | |
87 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
P12814 | |
ACTN1 | |
E. coli-derived recombinant human alpha-Actinin 1 Asp272-Thr500 Accession # P12814 | |
100 μg | |
Primary | |
Detects human, mouse and rat alpha-Actinin 1 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction