Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat alpha-Actinin 1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$227.50 - $549.50
Specifications
Antigen | alpha-Actinin 1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
alpha-Actinin 1 Polyclonal specifically detects alpha-Actinin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
alpha-Actinin 1 | |
Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Human, Mouse, Rat | |
P12814 | |
87 | |
E. coli-derived recombinant human alpha-Actinin 1 Asp272-Thr500 Accession # P12814 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human, mouse and rat alpha-Actinin 1 in Western blots. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
actinin 1 smooth muscle, actinin, alpha 1, ACTN1, alphaActinin 1, alpha-Actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, BDPLT15, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1 | |
ACTN1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title