Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat beta-2 Adrenergic R/ADRB2 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

$214.00 - $491.00

Specifications

Antigen beta-2 Adrenergic R/ADRB2
Clone 2204C
Dilution Western Blot 2 μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB10040SP
GREENER_CHOICE
View Documents
R&D Systems
MAB10040SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB10040100
GREENER_CHOICE
View Documents
R&D Systems
MAB10040100
100 μg
Each for $491.00
Only null left
Add to Cart
 
Description

Description

beta-2 Adrenergic R/ADRB2 Monoclonal specifically detects beta-2 Adrenergic R/ADRB2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

beta-2 Adrenergic R/ADRB2
Western Blot 2 μg/mL
Monoclonal
Supernatant
Human, Mouse, Rat
P07550
154.0
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
2204C
Western Blot
Unconjugated
Rabbit
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
ADRB2, ADRB2R, ADRBR, adrenergic, beta-2-, receptor, surface, B2AR, B2ARADRB2R, BAR, beta-2 adrenergic receptor, beta2 AdrenergicR, beta-2 AdrenergicR, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
Synthetic peptide containing human beta-2 Adrenergic R/ADRB2, Accession No. P07550
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human beta-2 Adrenergic R/ADRB2 in direct ELISAs. Detects human, mouse, and rat beta-2 Adrenergic R/ADRB2 in Western blots.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.