Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat GABAB R2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $351.50
Specifications
Antigen | GABA-B R2 |
---|---|
Clone | 2605C |
Dilution | Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10456SP
![]() |
R&D Systems
MAB10456SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10456100
![]() |
R&D Systems
MAB10456100 |
100 μg |
Each for $351.50
|
|
|||||
Description
GABA-B R2 Monoclonal antibody specifically detects GABA-B R2 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, ImmunohistochemistrySpecifications
GABA-B R2 | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
G protein-coupled receptor 51, GABABR2, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, GABBR2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, gb2, GPR51, GPR51GABA-B receptor, R2 subunit, GPRC3B, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099 | |
Mouse myeloma cell line NS0-derived recombinant rat GABAB R2, Trp41-Ser482, Accession # O88871 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2605C | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O88871 | |
9568 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title