Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat/Hamster ACE-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3437
This item is not returnable.
View return policy
Description
ACE-2 Monoclonal specifically detects ACE-2 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot.Specifications
ACE-2 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL | |
Q8R0I0 | |
ACE2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse ACE-2 Gln18-Thr740 (predicted) Accession # Q8R0I0 | |
100 μg | |
Primary | |
Detects mouse ACE-2 in direct ELISAs and human, mouse, rat and hamster ACE-2 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse ACE-1 is observed. | |
Human, Mouse, Rat, Hamster | |
Purified |
Western Blot | |
460502 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
59272 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction