Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat/Hamster ACE-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | ACE-2 |
---|---|
Clone | 460502 |
Concentration | LYOPH |
Dilution | Western Blot 0.5 ug/mL |
Applications | Western Blot |
Description
ACE-2 Monoclonal specifically detects ACE-2 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot.Specifications
ACE-2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse, Rat, Hamster | |
Q8R0I0 | |
59272 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse ACE-2 Gln18-Thr740 (predicted) Accession # Q8R0I0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse ACE-2 in direct ELISAs and human, mouse, rat and hamster ACE-2 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse ACE-1 is observed. |
460502 | |
Western Blot 0.5 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACE2, ACEH, ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
ACE2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title