Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Niemann-Pick Type C1/NPC1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | Niemann-Pick C1 |
---|---|
Clone | 1318A |
Dilution | Western Blot 0.5 μg/mL, Immunohistochemistry 3-25 μg/mL, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, Immunoassay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10105SP
![]() |
R&D Systems
MAB10105SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10105100
![]() |
R&D Systems
MAB10105100 |
100 μg |
Each for $491.00
|
|
|||||
Description
Niemann-Pick C1 Monoclonal specifically detects Niemann-Pick C1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.Specifications
Niemann-Pick C1 | |
Western Blot 0.5 μg/mL, Immunohistochemistry 3-25 μg/mL, Knockout Validated | |
Monoclonal | |
Supernatant | |
Human, Mouse, Rat | |
FLJ98532, Niemann-Pick C1 protein, Niemann-Pick disease, type C1, NPC, NPC1, SLC65A1 | |
Synthetic peptide containing Human Niemann-Pick Type C1/NPC1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Niemann-Pick Type C1/NPC1 in direct ELISAs. Detects human, mouse, and rat Niemann-Pick Type C1/NPC1 in Western blots. |
1318A | |
Western Blot, Immunohistochemistry, Immunoassay | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
4864 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title