Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat STAT5b Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB15841
This item is not returnable.
View return policy
Description
STAT5b Monoclonal specifically detects STAT5b in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.Specifications
STAT5b | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
6777 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Western Blot, Immunoprecipitation, Immunocytochemistry, Immunoassay | |
1032E | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunoprecipitation 2 ug/1 mg cell lysate, Immunocytochemistry 5-25 ug/mL, Knockout Validated | |
P51692 | |
STAT5B | |
Human STAT5b synthetic peptide aa 777-787 Accession # P51692 | |
100 μg | |
Primary | |
Detects human, mouse, and rat STAT5b in Western blots. | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction