Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat STAT5b Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $514.50
Specifications
Antigen | STAT5b |
---|---|
Clone | 1032E |
Dilution | Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunoprecipitation 2 ug/1 mg cell lysate, Immunocytochemistry 5-25 ug/mL, Knockout Validated |
Applications | Western Blot, Immunoprecipitation, Immunocytochemistry, Immunoassay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB15841SP
![]() |
R&D Systems
MAB15841SP |
25 μg |
Each for $214.00
|
|
|||||
MAB15841
![]() |
R&D Systems
MAB15841 |
100 μg |
Each for $514.50
|
|
|||||
Description
STAT5b Monoclonal specifically detects STAT5b in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.Specifications
STAT5b | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunoprecipitation 2 ug/1 mg cell lysate, Immunocytochemistry 5-25 ug/mL, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
signal transducer and activator of transcription 5B, STAT5, transcription factor STAT5B | |
STAT5B | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1032E | |
Western Blot, Immunoprecipitation, Immunocytochemistry, Immunoassay | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P51692 | |
6777 | |
Human STAT5b synthetic peptide aa 777-787 Accession # P51692 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human, mouse, and rat STAT5b in Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title