Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse sFRP-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2350.00
Specifications
Antigen | sFRP-3/FRZB |
---|---|
Clone | 133912 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
sFRP-3/FRZB Monoclonal specifically detects sFRP-3/FRZB in Human, Mouse samples. It is validated for Western Blot.Specifications
sFRP-3/FRZB | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
AAB51298 | |
2487 | |
Mouse myeloma cell line NS0-derived recombinant human sFRP-3 Ala33-Asn325 Accession # AAB51298 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human sFRP-3 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse (rm) sFRP-3 and no cross-reactivity with recombinant human (rh) sFRP-1, rmsFRP-2, or rhsFRP-4. |
133912 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FIZ, FREOS1, Frezzled, Fritz, frizzled-related protein, Frizzled-related protein 1, FRP, FRP-3, FRZB, Frzb1, FrzB-1, FRZB1sFRP-3, FRZB-PEN, FZRB, hFIZ, secreted frizzled-related protein 3, sFRP3, SFRP3frezzled, SRFP3frizzled homolog-related | |
FRZB | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title