Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse SR140 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
| Antigen | SR140 |
|---|---|
| Clone | 983615 |
| Dilution | Western Blot 2μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9897-SP
![]() |
R&D Systems
MAB9897SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9897-100
![]() |
R&D Systems
MAB9897100 |
100 μg |
Each for $468.00
|
|
|||||
Description
SR140 Monoclonal specifically detects SR140 in Human, Mouse samples. It is validated for Western Blot.Specifications
| SR140 | |
| Western Blot 2μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human, Mouse | |
| O15042 | |
| 23350 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 983615 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| 140 kDa Ser/Arg-rich domain protein, fSAPa, KIAA0332, SR140, U2 snRNP-associated SURP domain containing, U2 snRNP-associated SURP motif-containing protein, U2-associated protein SR140, U2-Associated SR140 Protein, U2SURP | |
| Synthetic peptide containing human SR140 Peptide Accession # O15042 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human SR140 in direct ELISAs. Detects human and mouse SR140 in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title