Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse STAT5a/b Pan Specific Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2168
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2168 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2168 Supplier R&D Systems Supplier No. AF2168
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 13 publications

STAT5a/b Polyclonal specifically detects STAT5a/b in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation (ChIP).

Specifications

Antigen STAT5a/b
Applications ChIP Assay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 20 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells
Gene Alias MGF, STAT5, STAT5A signal transducer and activator of transcription 5A
Gene Symbols STAT5A
Host Species Rabbit
Immunogen Human STAT5a and STAT5b synthetic peptide aa 219-390
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6776
Test Specificity Detects endogenous human and mouse STAT5A and STAT5B in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.