Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Sulfamidase/SGSH Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | Sulfamidase/SGSH |
---|---|
Clone | 1018331 |
Dilution | Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB83801SP
![]() |
R&D Systems
MAB83801SP |
25 μg |
Each for $200.50
|
|
|||||
MAB83801100
![]() |
R&D Systems
MAB83801100 |
100 μg |
Each for $432.50
|
|
|||||
Description
Sulfamidase/SGSH Monoclonal specifically detects Sulfamidase/SGSH in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Sulfamidase/SGSH | |
Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
heparan sulfate sulfatase, HSS, HSSN-sulphoglucosamine sulphohydrolase, MPS3A, MPS3Asulphamidase, N-sulfoglucosamine sulfohydrolase, SFMD, SFMDEC 3.10.1.1, SGSH, Sulfoglucosamine sulfamidase, Sulphamidase | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Sulfamidase/SGSH, Arg23-Leu502, Accession # P51688 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1018331 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P51688 | |
6448 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title