Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Syntaxin 12 Antibody, R&D Systems™

Goat Antibody
$200.50 - $468.50
Specifications
Antigen | Syntaxin 12 |
---|---|
Dilution | Immunohistochemistry 1-15 μg/mL |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Syntaxin 12 specifically detects Syntaxin 12 in Human, Mouse samples. It is validated for Immunohistochemistry.Specifications
Syntaxin 12 | |
Immunohistochemistry | |
Unconjugated | |
Goat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
MGC51957, STX12, STX13, STX14, syntaxin 12, syntaxin-12 | |
E.coli-derived recombinant human Syntaxin 12, Met1-Gln200, Accession No. Q86Y82 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Syntaxin 12 in direct ELISAs. Detects human and mouse Syntaxin 12 in immunohistochemistry. |
Immunohistochemistry 1-15 μg/mL | |
Polyclonal | |
Lyophilized | |
Human, Mouse | |
Q86Y82 | |
23673.0 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title