Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse TIMP-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF974
This item is not returnable.
View return policy
Description
TIMP-4 Polyclonal specifically detects TIMP-4 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
TIMP-4 | |
Polyclonal | |
Unconjugated | |
Q99727 | |
TIMP4 | |
Mouse myeloma cell line NS0-derived recombinant human TIMP-4 Cys30-Pro224 Accession # Q99727 | |
100 μg | |
Primary | |
Detects human and mouse TIMP-4 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) TIMP-1, rhTIMP-2, and rhTIMP-3 is observed. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4 | |
Goat | |
Affinity Purified | |
RUO | |
7079 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction