Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse TIMP-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | TIMP-4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
TIMP-4 Polyclonal specifically detects TIMP-4 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
TIMP-4 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human, Mouse | |
metalloproteinase inhibitor 4, TIMP metallopeptidase inhibitor 4, TIMP4, TIMP-4, tissue inhibitor of metalloproteinase 4, Tissue inhibitor of metalloproteinases 4 | |
TIMP4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q99727 | |
7079 | |
Mouse myeloma cell line NS0-derived recombinant human TIMP-4 Cys30-Pro224 Accession # Q99727 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human and mouse TIMP-4 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) TIMP-1, rhTIMP-2, and rhTIMP-3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title