Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse VEGF-B186 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | VEGF-B |
---|---|
Clone | 67409 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
VEGF-B Monoclonal specifically detects VEGF-B in Human, Mouse samples. It is validated for Western Blot.Specifications
VEGF-B | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL | |
VEGFB | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
67409 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P49765 | |
7423 | |
E. coli-derived recombinant human VEGF-B 186 Accession # P49765 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human VEGF-B 186 in direct ELISAs and Western blots. In these formats, this antibody shows 100% cross-reactivity with recombinant mouse (rm) VEGF-B 186 but does not detect rh or rmVEGF-B 167 rhVEGF 206 rhVEGF 167 rhVEGF-C, rhVEGF-D, rhCTGF, rhPlGF, or rhLDGF. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title