Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Wnt-3a Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 2 publications

$241.00 - $549.50

Specifications

Antigen Wnt-3a
Clone 217804.2R
Dilution Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/mL, CyTOF-ready
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB13242SP
GREENER_CHOICE
View Documents
R&D Systems
MAB13242SP
25 μg
Each for $241.00
Only null left
Add to Cart
 
MAB13242100
GREENER_CHOICE
View Documents
R&D Systems
MAB13242100
100 μg
Each for $549.50
Only null left
Add to Cart
 
Description

Description

Wnt-3a Monoclonal specifically detects Wnt-3a in Human, Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications

Specifications

Wnt-3a
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/mL, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
MGC119418, MGC119419, MGC119420, protein Wnt-3a, wingless-type MMTV integration site family, member 3A, Wnt3a
E.coli-derived recombinant mouse Wnt-3a Ser36-Gln75, Trp219-Arg269 Accession # P27467
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Wnt-3a in direct ELISAs.
217804.2R
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Rat
Human, Mouse
P27487
89780
IgG2a
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.