Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MSPR/Ron Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $573.00
Specifications
Antigen | MSPR/Ron |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 2 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Blockade of Receptor-ligand Interaction 1-3 ug/mL |
Applications | Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
MSPR/Ron Polyclonal specifically detects MSPR/Ron in Human samples. It is validated for Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction.Specifications
MSPR/Ron | |
Western Blot 2 ug/mL, Immunoprecipitation 1 ug/1 mg cell lysate, Blockade of Receptor-ligand Interaction 1-3 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 | |
MST1R | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
CAA49634 | |
4486 | |
Mouse myeloma cell line NS0-derived recombinant human MSP R Glu25-Ser956 Accession # CAA49634 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Delects human MSP R/Ron in direct ELISAs and Western blots. In these formats, approximately 25% cross-reactivity with recombinant mouse MSP R is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title