Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NF-M Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $514.50
Specifications
Antigen | NF-M |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1-2 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, KnockDown |
Classification | Polyclonal |
Description
NF-M Polyclonal specifically detects NF-M in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.Specifications
NF-M | |
Western Blot 1-2 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated | |
Polyclonal | |
Sheep | |
Human | |
NEF3, NEF3neurofilament medium polypeptide, NEFM, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NFM, NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein | |
NEFM | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, KnockDown | |
Unconjugated | |
RUO | |
P07197 | |
4741 | |
E. coli-derived recombinant human NF-M Ser2-Ser358 Accession # P07197 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human NF-M in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) NF-H and rhNF-L is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title