Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NT-3 Propeptide Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $514.50
Specifications
Antigen | NT-3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
NT-3 Polyclonal specifically detects NT-3 in Human samples. It is validated for Western Blot.Specifications
NT-3 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3, NTF3 | |
NTF3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P20783 | |
4908 | |
E. coli-derived recombinant human NT-3 Asn19-Arg138 Accession # P20783 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human NT-3 Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant mouse NT-3 Propeptide is observed, 2% cross-reactivity with recombinant human NT-4 propeptide is observed, and less than 1% cross-reactivity with recombinant human NT-3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title