Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human NT-4 DuoSet ELISA
Click to view available options
Quantity:
15 x 96-well Plates
Description
The Human NT-4 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human NT-4.
Specifications
Specifications
Quantity | 15 x 96-well Plates |
Conjugate | Biotin |
Assay Range | 31.20 to 2000 pg/mL |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 4909 |
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction