Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Olig 1, 2, 3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Olig1/2/3 |
---|---|
Clone | 257224 |
Concentration | LYOPH |
Dilution | Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Description
Olig1/2/3 Monoclonal specifically detects Olig1/2/3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
Olig1/2/3 | |
LYOPH | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
BHLHB1, bHLHb6, BHLHB7, bHLHe21, PRKCBP2, RACK17 | |
E. coli-derived recombinant human Olig1 aa 1-272 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Olig 1, 2, 3. In direct ELISAs, this antibody shows approximately 20% cross-reactivity with recombinant human (rh) Olig2 and 50 - 100% cross-reactivity with rhOlig3. |
257224 | |
Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
116448 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title