Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human p53 ELISA Kit (Colorimetric)
SDP

Catalog No. nbp191257 Shop All R&D Systems Products
Click to view available options
Quantity:
96 Tests

Catalog No. NBP191257

Supplier: Novus Biologicals™ NBP191257

Only null left
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human p53 ELISA Kit (Colorimetric) measures human p53 in cell culture supernatant, serum.

Specifications

Assay Range 0.78 - 50 U/mL
Assay Sensitivity 0.33 U/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 7157
Gene Symbol TP53
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human p53, Biotin-Conjugate anti-human p53 monoclonal antibody, Streptavidin-HRP, Human p53 Standard lyophilized, 100U/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Stem Cell Markers, Transcripti
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
Target p53
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.