Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Parechovirus 1/3 VP1 Antibody (HL2427) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295103 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295103 Supplier Novus Biologicals Supplier No. NBP325514
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Human Parechovirus 1/3 VP1 Monoclonal antibody specifically detects Human Parechovirus 1/3 VP1 in Virus samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Human Parechovirus 1/3 VP1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HL2427
Conjugate Unconjugated
Dilution Western Blot 1:500-1:10000, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias Capsid protein VP1, Human Parechovirus Capsid protein VP1, P1D, Parechovirus A VP1, PeV-A VP1, Virion protein 1
Host Species Rabbit
Immunogen Full length of Human Parechovirus 3 VP1 recombinant protein.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.