Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDGF-B Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2020.00
Specifications
Antigen | PDGF-B |
---|---|
Clone | 108132 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot.Specifications
PDGF-B | |
Western Blot 1 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P01127 | |
5155 | |
E. coli-derived recombinant human PDGF-BB Ser82-Thr190 Accession # P01127 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human PDGF-B subunit in direct ELISAs and Western blots. In direct ELISAs, shows approximately 40% cross-reactivity with recombinant human (rh) PDGF-AB and no cross-reactivity with rhCTGF, rhLDGF, rhPDGF-AA, recombinant rat (rr) PDGF-AA, rrPDGF-AB, rrPDGF-BB, rhPDGF-C, rhPDGF-D, rhPlGF, rhVEGF, rhVEGF-C, or rhVEGF-D. |
108132 | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B chain, Becaplermin, C-Sis, FLJ12858, IBGC5, PDGF subunit B, PDGF2, PDGF-2, PDGFB, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SIS, SSV | |
PDGFB | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title