Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PDGF-B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2201SP
This item is not returnable.
View return policy
Description
PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot.Specifications
PDGF-B | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B chain, Becaplermin, C-Sis, FLJ12858, IBGC5, PDGF subunit B, PDGF2, PDGF-2, PDGFB, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SIS, SSV | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
5155 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot | |
108104 | |
Western Blot 1 ug/mL | |
P01127 | |
PDGFB | |
E. coli-derived recombinant human PDGF-BB Ser82-Thr190 Accession # P01127 | |
25 μg | |
Primary | |
Detects human PDGF-B subunit in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 100% cross-reactivity with recombinant human (rh) PDGF-AB and recombinant rat PDGF-BB, 15% cross-reactivity with rhPDGF-AA, and no cross-reactivity with rhPlGF, rhVEGF, rhVEGF-C, rhVEGF-D, rhCTGF, or rhLDGF. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction