Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PDGF-B Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $1798.00

Specifications

Antigen PDGF-B
Clone 108104
Dilution Western Blot 1 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB2201SP
GREENER_CHOICE
View Documents
R&D Systems
MAB2201SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB2201100
GREENER_CHOICE
View Documents
R&D Systems
MAB2201100
100 μg
Each for $479.50
Only null left
Add to Cart
 
MAB2201500
GREENER_CHOICE
View Documents
R&D Systems
MAB2201500
500 μg
Each for $1,798.00
Only null left
Add to Cart
 
Description

Description

PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot.
Specifications

Specifications

PDGF-B
Western Blot 1 ug/mL
Monoclonal
Mouse
Human
P01127
5155
E. coli-derived recombinant human PDGF-BB Ser82-Thr190 Accession # P01127
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human PDGF-B subunit in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 100% cross-reactivity with recombinant human (rh) PDGF-AB and recombinant rat PDGF-BB, 15% cross-reactivity with rhPDGF-AA, and no cross-reactivity with rhPlGF, rhVEGF, rhVEGF-C, rhVEGF-D, rhCTGF, or rhLDGF.
108104
Western Blot
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
B chain, Becaplermin, C-Sis, FLJ12858, IBGC5, PDGF subunit B, PDGF2, PDGF-2, PDGFB, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SIS, SSV
PDGFB
IgG2b
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.