Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Pepsinogen A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7525SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7525SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7525SP Supplier R&D Systems Supplier No. MAB7525SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pepsinogen A Monoclonal specifically detects Pepsinogen A in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Pepsinogen A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 762015
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P00790
Gene Alias EC 3.4.23, EC 3.4.23.1, FLJ58952, FLJ77962, pepsin A, pepsinogen 4, group I (pepsinogen A), pepsinogen A4, PGA3, PGA5
Gene Symbols PGA4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Pepsinogen A4/PGA4 Ile16-Ala388 Accession # P00790
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643847
Test Specificity Detects human Pepsinogen A4/PGA4 in direct ELISAs. Human PGA3 and PGA5 reactivity has not been demonstrated but is likely due to their high homology with human PGA4. In direct ELISAs, no cross-reactivity with recombinant human (rh) Pepsinogen C, rhCathepsin D, or rhCathepsin E is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.