Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Human PGLYRP4/PGRP-I beta Antibody, R&D Systems™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
| Antigen | PGLYRP4/PGRP-I beta | 
|---|---|
| Concentration | LYOPH | 
| Dilution | Western Blot 0.1 ug/mL | 
| Applications | Western Blot | 
| Classification | Polyclonal | 
Description
PGLYRP4/PGRP-I beta Polyclonal specifically detects PGLYRP4/PGRP-I beta in Human samples. It is validated for Western Blot.Specifications
| PGLYRP4/PGRP-I beta | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| peptidoglycan recognition protein 4, peptidoglycan recognition protein I beta, Pglyrp-1 beta, PGRPI beta, PGRP-I beta, PGRP-I-beta | |
| PGLYRP4 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | 
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q96LB8 | |
| 57115 | |
| Mouse myeloma cell line NS0-derived recombinant human PGLYRP4/PGRP-I beta Asp18-Lys372 Accession # Q96LB8 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human PGLYRP4/PGRP-I beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse PGRP-I beta is observed and 10% cross-reactivity with recombinant human PGRP-S is observed. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title