Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Phospho-FGFR1 DuoSet IC ELISA
Click to view available options
Packaging Quantity:
15 Plates
2 Plates
5 Plates
Description
The Human Phospho-FGF R1 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human FGFR1.
Specifications
Specifications
Product Type | ELISA Development Kit for Phosphorylated Intracellular Proteins |
Quantity | 1 Kit |
Species | Human |
Conjugate | Biotin |
Assay Range | 125.00 to 8000 pg/mL |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 2260 |
Synonym | basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGF R1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, Flt-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2 |
Packaging Quantity | 15 Plates |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction