Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-Histone H2AX (S139) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$227.50 - $538.00
Specifications
Antigen | Histone H2AX |
---|---|
Clone | 2207D |
Dilution | Western Blot 0.25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB2288SP
![]() |
R&D Systems
MAB2288SP |
25 μg |
Each for $227.50
|
|
|||||
MAB2288100
![]() |
R&D Systems
MAB2288100 |
100 μg |
Each for $538.00
|
|
|||||
Description
Histone H2AX Monoclonal specifically detects Histone H2AX in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
Histone H2AX | |
Western Blot 0.25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Human | |
P16104 | |
3014 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
2207D | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
H2A.X, H2A/X, H2AFX | |
Phospho-peptide containing human Histone H2AX S139 site Accession # P16104 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Histone H2AX when phosphorylated at S139 in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title