Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-p53 (S392) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | p53 |
---|---|
Clone | 2185A |
Dilution | Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
MAB2996SP
![]() |
R&D Systems
MAB2996SP |
Unconjugated | 25 μg |
Each for $200.50
|
|
||||
MAB2996100
![]() |
R&D Systems
MAB2996100 |
Unconjugated | 100 μg |
Each for $410.00
|
|
||||
Description
p53 Monoclonal specifically detects p53 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
p53 | |
Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
Phosphopeptide containng the human p53 S392 site Accession # P04637 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human p53 when phoshorylated at S392 |
2185A | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P04637 | |
7157 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title