Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-Ribosomal Protein S6/RPS6 (S235/S236) Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Ribosomal Protein S6/RPS6 |
---|---|
Clone | 973210 |
Dilution | Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB54361SP
![]() |
R&D Systems
MAB54361SP |
25 μg |
Each for $200.50
|
|
|||||
MAB54361100
![]() |
R&D Systems
MAB54361100 |
100 μg |
Each for $502.50
|
|
|||||
Description
Ribosomal Protein S6/RPS6 Monoclonal specifically detects Ribosomal Protein S6/RPS6 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Ribosomal Protein S6/RPS6 | |
Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
P62753 | |
6194 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
973210 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, RPS6, S6 | |
Phosphopeptide containing human Ribosomal Protein S6 S235/S236 sites Accession # P62753 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Ribosomal Protein S6 when dually phosphorylated at S235 and S236. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title