Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PIN Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB877
This item is not returnable.
View return policy
Description
PIN/DLC8 Monoclonal specifically detects PIN/DLC8 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
PIN/DLC8 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cytoplasmic dynein light polypeptide, DLC1, DLC1DNCLC1, DLC8, DLC8MGC126137, DNCL1, DNCL1MGC126138, DNCLC1, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, DYNLL1, hdlc1, LC8, LC8a, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
8655 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunoprecipitation | |
98618 | |
Western Blot 1 ug/mL, Immunoprecipitation 2.5-5 ug/10^6 cells | |
P63167 | |
DYNLL1 | |
E. coli-derived recombinant human PIN Trp2-Gly89 Accession # P63167 | |
500 μg | |
Primary | |
Detects human PIN in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction