Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PIN Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | PIN/DLC8 |
---|---|
Clone | 98618 |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 2.5-5 ug/10^6 cells |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
PIN/DLC8 Monoclonal specifically detects PIN/DLC8 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
PIN/DLC8 | |
Western Blot 1 ug/mL, Immunoprecipitation 2.5-5 ug/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cytoplasmic dynein light polypeptide, DLC1, DLC1DNCLC1, DLC8, DLC8MGC126137, DNCL1, DNCL1MGC126138, DNCLC1, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, DYNLL1, hdlc1, LC8, LC8a, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase | |
DYNLL1 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
98618 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P63167 | |
8655 | |
E. coli-derived recombinant human PIN Trp2-Gly89 Accession # P63167 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human PIN in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title