Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human PIN Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7063997
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF877SP 25 μg
AF877 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF877SP Supplier R&D Systems Supplier No. AF877SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PIN/DLC8 Polyclonal specifically detects PIN/DLC8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen PIN/DLC8
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 2.5-5 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P63167
Gene Alias cytoplasmic dynein light polypeptide, DLC1, DLC1DNCLC1, DLC8, DLC8MGC126137, DNCL1, DNCL1MGC126138, DNCLC1, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, DYNLL1, hdlc1, LC8, LC8a, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase
Gene Symbols DYNLL1
Host Species Goat
Immunogen E. coli-derived recombinant human PIN Trp2-Gly89 Accession # P63167
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8655
Test Specificity Detects human PIN in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.