Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Podocalyxin Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Catalog No. RD10004725
Change view
Click to view available options
:
100 μg; Alexa Fluor 700
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
RD10004725 100 μg; Alexa Fluor 700
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10004725 Supplier R&D Systems Supplier No. AF1658N100UG
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Podocalyxin Like Polyclonal antibody specifically detects Podocalyxin Like in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Gene Knock-Out

Specifications

Antigen Podocalyxin Like
Applications Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Gene Knock-Out
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation Supplied 0.2 mg/mL in 1X PBS with RDF1 and 0.09% Sodium Azide
Gene Alias GCTM, Gp200MGC138240, PCLP, PCLP1, PCLP-1, PCLPGCTM-2 antigen, PCpodocalyxin, POD XL, podocalyxin-like, Podocalyxin-like protein 1, PODXL
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Podocalyxin, Ser23-Arg427, Accession # AAB61574
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5420
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8 °C as supplied
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.