Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PPAR alpha/NR1C1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | PPAR alpha/NR1C1 |
---|---|
Clone | 2297B |
Dilution | Immunocytochemistry 3-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB24781SP
![]() |
R&D Systems
MAB24781SP |
25 μg |
Each for $200.50
|
|
|||||
MAB24781100
![]() |
R&D Systems
MAB24781100 |
100 μg |
Each for $410.00
|
|
|||||
Description
PPAR alpha/NR1C1 Monoclonal specifically detects PPAR alpha/NR1C1 in Human samples. It is validated for Immunocytochemistry.Specifications
PPAR alpha/NR1C1 | |
Immunocytochemistry 3-25 μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
Q07869 | |
5465 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
2297B | |
Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
alpha, hPPAR, NR1C1, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARA, PPARalpha, PPAR-alpha, PPARMGC2452 | |
E. coli-derived recombinant human PPAR alpha/NR1C1 Met1-Asn99 Accession # Q07869 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human PPAR alpha/NR1C1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title