Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Primate BMP-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB7571100
This item is not returnable.
View return policy
Description
BMP-4 Monoclonal specifically detects BMP-4 in Human, Primate samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| BMP-4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 652 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, ELISA | |
| 66110 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P12644 | |
| BMP4 | |
| Mouse myeloma cell line NS0-derived recombinant human BMP-4 Ser293-Arg408 Accession # P12644 | |
| 100 μg | |
| Primary | |
| Detects human and primate BMP-4 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) BMP-2, rhBMP-3, rhBMP-5, rhBMP-6, or rhBMP-7 under non-reducing conditions. | |
| Human, Primate | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction