Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Primate BMP-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
$200.50 - $1973.00
Specifications
| Antigen | BMP-4 |
|---|---|
| Clone | 66110 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB7571SP
![]() |
R&D Systems
MAB7571SP |
25 μg |
Each for $200.50
|
|
|||||
MAB7571100
![]() |
R&D Systems
MAB7571100 |
100 μg |
Each for $538.00
|
|
|||||
MAB7571500
![]() |
R&D Systems
MAB7571500 |
500 μg |
Each for $1,973.00
|
|
|||||
Description
BMP-4 Monoclonal specifically detects BMP-4 in Human, Primate samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| BMP-4 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Primate | |
| P12644 | |
| 652 | |
| Mouse myeloma cell line NS0-derived recombinant human BMP-4 Ser293-Arg408 Accession # P12644 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and primate BMP-4 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) BMP-2, rhBMP-3, rhBMP-5, rhBMP-6, or rhBMP-7 under non-reducing conditions. |
| 66110 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME | |
| BMP4 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title