Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Primate VEGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB293R100
This item is not returnable.
View return policy
Description
VEGF Monoclonal specifically detects VEGF in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
VEGF | |
Monoclonal | |
Unconjugated | |
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
7422 | |
Human, Primate | |
Purified |
Western Blot, Immunohistochemistry, Neutralization, ELISA | |
26503R | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 10-60 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
NP_001165097 | |
VEGFA | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGF 165 Ala27-Arg191 Accession # NP_001165097 | |
100 μg | |
Primary | |
Detects human VEGF 165 and human VEGF 121 in direct ELISAs and Western blots. In ELISAs, this antibody shows approximately 10% cross-reactivity with recombinant mouse (rm) VEGF and rrVEGF and no cross-reactivity with rhVEGF-D. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction