Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Primate VEGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
$573.00
Specifications
Antigen | VEGF |
---|---|
Clone | 26503R |
Dilution | Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 10-60 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, Immunohistochemistry, Neutralization, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB293R100
![]() |
R&D Systems
MAB293R100 |
100 μg |
Each of 1 for $573.00
|
|
|||||
Description
VEGF Monoclonal specifically detects VEGF in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
VEGF | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 10-60 ng/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
NP_001165097 | |
7422 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Detects human VEGF 165 and human VEGF 121 in direct ELISAs and Western blots. In ELISAs, this antibody shows approximately 10% cross-reactivity with recombinant mouse (rm) VEGF and rrVEGF and no cross-reactivity with rhVEGF-D. |
26503R | |
Western Blot, Immunohistochemistry, Neutralization, ELISA | |
Unconjugated | |
Mouse | |
Human, Primate | |
MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
VEGFA | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title