Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human R-Spondin 1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $561.00
Specifications
Antigen | R-Spondin 1 |
---|---|
Concentration | LYOPH |
Dilution | Neutralization 3-12 ug/mL |
Applications | Neutralization |
Classification | Polyclonal |
Description
R-Spondin 1 Polyclonal specifically detects R-Spondin 1 in Human samples. It is validated for Neutralization.Specifications
R-Spondin 1 | |
Neutralization 3-12 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
Cristin 3, CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPO1, RSPONDIN, RSpondin 1, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1 | |
RSPO1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Neutralization | |
Unconjugated | |
RUO | |
Q2MKA7 | |
284654 | |
Chinese hamster ovary cell line CHO-derived human R-Spondin 1 Arg31-Ala263 Accession # Q2MKA7 | |
Primary | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Detects human R-Spondin 1 in direct ELISAs. In direct ELISAs, approximately 6% cross-reactivity with recombinant mouse R-Spondin 1 is observed and less than 1% cross-reactivity with recombinant human (rh) R-Spondin 2, rhR-Spondin 3, and rhR-Spondin 4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title