Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Rat Annexin V Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7112313
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB399SP 25 μg
MAB399100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB399SP Supplier R&D Systems Supplier No. MAB399SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Annexin V Monoclonal specifically detects Annexin V in Human, Rat samples. It is validated for Western Blot, Simple Western, Knockout Validated.

Specifications

Antigen Annexin V
Applications Western Blot
Classification Monoclonal
Clone 39103
Conjugate Unconjugated
Dilution Western Blot 0.25 ug/mL, Simple Western 2.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P08758
Gene Alias Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, ANXA5, ANXV, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha
Host Species Mouse
Immunogen E. coli-derived recombinant human Annexin V Met1-Asp320 Accession # P08758
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308
Test Specificity Detects human Annexin V in direct ELISAs and Western blots. Detects rat Annexin V in Simple Westerns TM .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.