Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat CCR4 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Supplier: R&D Systems FAB1567R025
This item is not returnable.
View return policy
Description
CCR4 Monoclonal specifically detects CCR4 in Human, Rat samples. It is validated for Flow Cytometry.Specifications
CCR4 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1233 | |
Human, Rat | |
Supernatant |
Flow Cytometry | |
205410 | |
Flow Cytometry 5 uL/10^6 cells | |
P51679 | |
CCR4 | |
Human CCR4 transfectants Met1-Leu360 Accession # P51679 | |
25 Tests | |
Primary | |
Stains human CCR4-transfected cell lines but not the corresponding parent cell lines. Also detects human CCR4 on human PBLs and platelets and rat CCR4 on rat splenocytes. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction