Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat CCR4 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
$186.30
Specifications
Antigen | CCR4 |
---|---|
Clone | 205410 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1567R025
![]() |
R&D Systems
FAB1567R025 |
25 Tests |
Each of 1 for $186.30
|
|
|||||
Description
CCR4 Monoclonal specifically detects CCR4 in Human, Rat samples. It is validated for Flow Cytometry.Specifications
CCR4 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Supernatant | |
RUO | |
P51679 | |
1233 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Stains human CCR4-transfected cell lines but not the corresponding parent cell lines. Also detects human CCR4 on human PBLs and platelets and rat CCR4 on rat splenocytes. |
205410 | |
Flow Cytometry | |
Alexa Fluor 647 | |
Mouse | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
CCR4 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title