Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Rat IL-32 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | IL-32 |
---|---|
Clone | 2525B |
Dilution | Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB6769SP
![]() |
R&D Systems
MAB6769SP |
25 μg |
Each for $200.50
|
|
|||||
MAB6769100
![]() |
R&D Systems
MAB6769100 |
100 μg |
Each for $479.50
|
|
|||||
Description
IL-32 Monoclonal specifically detects IL-32 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
IL-32 | |
Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
IL32, IL-32, IL-32alpha, IL-32beta, interleukin 32, interleukin-32, interleukin-32 theta, natural killer cell transcript 4, Natural killer cells protein 4, NK4, NK4IL-32delta, TAIF, TAIFa, TAIFb, TAIFc, TAIFd, TAIFIL-32gamma, Tumor necrosis factor alpha-inducing factor | |
E. coli-derived human IL-32 beta protein, Met1-Lys188, Accession # NP_001012649 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IL-32 beta and other IL-32 isoforms in direct ELISAs. In direct ELISAs, 95% cross-reactivity with recombinant human (rh) IL-32 alpha and 45% cross-reactivity with rhIL-32 gamma is detected. |
2525B | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat | |
NP_001012649 | |
9235 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title