Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Resistin ELISA Kit (Colorimetric)
SDP

Catalog No. nbp183744 Shop All Novus Biologicals Products
Click to view available options
Quantity:
96 Tests

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human Resistin ELISA Kit (Colorimetric) measures human Resistin in cell culture supernatant, serum, plasma (edta, citrate, heparin).

Specifications

Assay Range 31 - 2,000pg/mL
Assay Sensitivity 3.1pg/mL
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 56729
Gene Symbol RETN
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to human Resistin, Biotin-Conjugate anti-human Resistin polyclonal antibody, Streptavidin-HRP, Human Resistin Standard lyophilized, 4000 pg/ml upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Diabetes Research
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Adipose tissue-specific secretory factor, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-
Target Resistin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.