Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Respiratory Syncytial Virus A2 Fusion Protein Antibody (302)
SDP

Catalog No. NB020997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020997 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB020997 Supplier Novus Biologicals Supplier No. NBP289760

Rabbit Monoclonal Antibody

Human Respiratory Syncytial Virus A2 Fusion Protein Monoclonal antibody specifically detects Human Respiratory Syncytial Virus A2 Fusion Protein in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Human Respiratory Syncytial Virus A2 Fusion Protein
Applications Western Blot, ELISA
Classification Monoclonal
Clone 302
Conjugate Unconjugated
Dilution Western Blot, Sandwich ELISA Detection 1:1000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias HRSVgp08, RSV, RSV fusion protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Respiratory Syncytial Virus A2 Fusion Protein (AAB59858.1; Met1-Thr529).
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Has cross-reactivity in ELISA and WB with: Human (RSV) (A2) RSV-F Protein Human (RSV) RSV-F (Strain RSS-2) Protein Human (RSV) Fusion Protein) Human (RSV) Fusion Protein
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.